(R)-2-Amino-2-phenylacetamide ≥95%

Supplier: Matrix Scientific

Synonyms: D(-)-Phenylglycinamide, H-D-Phg-NH₂·HCl, H-D-Phg-NH2

076023-100G 076023-25G
102544-148EA 376.92 USD
102544-148 102833-898
(R)-2-Amino-2-phenylacetamide ≥95%
(R)-2-Amino-2-phenylacetamide

Hazard: Irritant/Refrigerate

Formula: C₈H₁₀N₂O
MW: 150.181 g/mol
Melting Pt: 125…129 °C
MDL Number: MFCD06799064
CAS Number: 6485-67-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR