1-(Aminomethyl)cyclopentanol hydrochloride ≥95%

Supplier: Matrix Scientific

076022-1G 076022-5G
102544-140EA 243.08 USD
102544-140 102544-142
1-(Aminomethyl)cyclopentanol hydrochloride ≥95%
1-(Aminomethyl)cyclopentanol hydrochloride

Hazard: Irritant

Formula: C₆H₁₄ClNO
MW: 151.64 g/mol
MDL Number: MFCD10686694
CAS Number: 76066-27-8

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR