tert-Butyl bis(2-bromoethyl)carbamate ≥95%

Supplier: Matrix Scientific

076017-250MG 076017-1G
102544-114EA 121.54 USD
102544-114 102544-116
tert-Butyl bis(2-bromoethyl)carbamate ≥95%
tert-Butyl bis(2-bromoethyl)carbamate

Hazard: Irritant

Formula: C₉H₁₇Br₂NO₂
MDL Number: MFCD11519276
CAS Number: 159635-50-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR