2-Amino-2'-fluoroacetophenone hydrochloride ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Fluorophenacylamine hydrochloride, 2-Amino-1-(2-fluorophenyl)ethanone hydrochloride

076016-1G 076016-5G
102544-110EA 569.23 USD
102544-110 102544-112
2-Amino-2'-fluoroacetophenone hydrochloride ≥95%
2-Amino-2'-fluoroacetophenone hydrochloride

Hazard: Irritant

Formula: C₈H₉ClFNO
MDL Number: MFCD08532458
CAS Number: 93102-96-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR