N-Methyldicyclohexylamine ≥95%
Supplier: Matrix Scientific
076013-100G
076013-500G
102544-100EA
78.46
USD
102544-100
102552-000
N-Methyldicyclohexylamine ≥95%
N-Methyldicyclohexylamine
Hazard: Irritant
Formula:
C₁₃H₂₅N |
MDL Number:
MFCD00014289 CAS Number: 7560-83-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...