(1S,4S)-2-Benzyl-2,5-diazabicyclo[2.2.1]heptane dihydrobromide ≥95%

Supplier: Matrix Scientific

076010-1G 076010-5G 076010-10G
102544-084EA 249.23 USD
102544-084 102544-086 102544-088
(1S,4S)-2-Benzyl-2,5-diazabicyclo[2.2.1]heptane dihydrobromide ≥95%
(1S,4S)-2-Benzyl-2,5-diazabicyclo[2.2.1]heptane dihydrobromide

Hazard: Irritant

Formula: C₁₂H₁₆N₂
MW: 350.10 g/mol
MDL Number: MFCD01321292
CAS Number: 116258-17-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR