2-Amino-3-(trifluoromethoxy)benzoic acid ≥95%

Supplier: Matrix Scientific

076008-1G 076008-5G 076008-25G
102544-072EA 60 USD
102544-072 102544-074 102833-896
2-Amino-3-(trifluoromethoxy)benzoic acid ≥95%
2-Amino-3-(trifluoromethoxy)benzoic acid

Hazard: Irritant

Formula: C₈H₆F₃NO₃
MDL Number: MFCD08669644
CAS Number: 561304-41-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR