3,5-Dimethylbenzyl bromide ≥95%

Supplier: Matrix Scientific

076005-5G 076005-100G
102544-056EA 44.62 USD
102544-056 102544-060
3,5-Dimethylbenzyl bromide ≥95%
3,5-Dimethylbenzyl bromide

Hazard: Corrosive/Lachrymator

Formula: C₉H₁₁Br
Melting Pt: 36…40 °C
MDL Number: MFCD00013539
CAS Number: 27129-86-8
UN: 3261
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR