Triclabendazole ≥95%

Supplier: Matrix Scientific

Synonyms: 6-Chloro-5-(2,3-dichlorophenoxy)-2-(methylthio)-1H-benzo[d]imidazole, 5-Chloro-6-(2,3-dichlorophenoxy)-2-(methylthio)benzimidazole, 6-Chloro-5-(2,3-dichlorophenoxy)-2-(methylthio)benzimidazole

076003-10G 076003-100G
102544-048EA 107.69 USD
102544-048 102544-050
Triclabendazole ≥95%
Triclabendazole

Hazard: Irritant

Formula: C₁₄H₉Cl₃N₂OS
MW: 359.66 g/mol
MDL Number: MFCD00864519
CAS Number: 68786-66-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR