8-Hydroxyquinoline-N-Oxide ≥95%

Supplier: Matrix Scientific

Synonyms: 8-Quinolinol N-Oxide, 8-Hydroxyquinoline 1-oxide

076002-10G 076002-100G
102544-044EA 160 USD
102544-044 102544-046
8-Hydroxyquinoline-N-Oxide ≥95%
8-Hydroxyquinoline-N-Oxide

Hazard: Irritant

Formula: C₉H₇NO₂
MW: 161.16 g/mol
MDL Number: MFCD00006739
CAS Number: 1127-45-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR