3-Phenylpropiolaldehyde ≥95%

Supplier: Matrix Scientific

Synonyms: Phenylpropargyl aldehyde

076000-1G 076000-5G 076000-10G
102544-032EA 69.23 USD
102544-032 102544-034 102544-036
3-Phenylpropiolaldehyde ≥95%
3-Phenylpropiolaldehyde

Hazard: Irritant

Formula: C₉H₆O
MW: 130.15 g/mol
MDL Number: MFCD00006995
CAS Number: 2579-22-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR