5-Fluoro-2-methylpyrimidin-4(3H)-one ≥95%

Supplier: Matrix Scientific

Synonyms: 5-Fluoro-2-methylpyrimidin-4-ol

075999-250MG 075999-1G 075999-5G
102544-026EA 456.92 USD
102544-026 102544-028 102544-030
5-Fluoro-2-methylpyrimidin-4(3H)-one ≥95%
5-Fluoro-2-methylpyrimidin-4(3H)-one

Hazard: Irritant

Formula: C₅H₅FN₂O
MW: 128.11 g/mol
MDL Number: MFCD16988103
CAS Number: 1480-91-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR