Ethyl-2-amino-4,5-dichlorobenzoate ≥95%

Supplier: Matrix Scientific

075998-250MG 075998-1G 075998-5G
102544-020EA 456.92 USD
102544-020 102544-022 102544-024
Ethyl-2-amino-4,5-dichlorobenzoate ≥95%
Ethyl 2-amino-4,5-dichlorobenzoate

Hazard: Irritant

Formula: C₉H₉Cl₂NO₂
MDL Number: MFCD10566266
CAS Number: 1108668-25-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR