Ethyl pyrrolidine-3-carboxylate hydrochloride ≥95%

Supplier: Matrix Scientific

Synonyms: Pyrrolidine-3-carboxylic acid ethyl ester hydrochloride

075995-1G 075995-5G
102544-004EA 64.62 USD
102544-004 102544-006
Ethyl pyrrolidine-3-carboxylate hydrochloride ≥95%
Ethyl pyrrolidine-3-carboxylate hydrochloride

Hazard: Irritant

Formula: C₇H₁₄ClNO₂
MW: 179.65 g/mol
MDL Number: MFCD14585012
CAS Number: 80028-44-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR