1-Bromobutane-2,3-dione ≥95%

Supplier: Matrix Scientific

075989-1G 075989-5G 075989-25G
102543-972EA 176.92 USD
102543-972 102543-974 102833-894
1-Bromobutane-2,3-dione ≥95%
1-Bromobutane-2,3-dione

Hazard: Irritant/Refrigerate

Formula: C₄H₅BrO₂
CAS Number: 5308-51-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR