Ethyl-2-amino-4-trifluoromethylbenzoate ≥95%

Supplier: Matrix Scientific

075988-1G 075988-5G
102543-968EA 335.38 USD
102543-968 102543-970
Ethyl-2-amino-4-trifluoromethylbenzoate ≥95%
Ethyl-2-amino-4-trifluoromethylbenzoate

Hazard: Irritant

Formula: C₁₀H₁₀F₃NO₂
MDL Number: MFCD08274771
CAS Number: 65568-55-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR