Methyl morpholine-2-carboxylate 2,2,2-trifluoroacetate ≥95%

Supplier: Matrix Scientific

075984-1G 075984-5G
102543-948EA 463.08 USD
102543-948 102543-950
Methyl morpholine-2-carboxylate 2,2,2-trifluoroacetate ≥95%
Methyl morpholine-2-carboxylate 2,2,2-trifluoroacetate

Hazard: Irritant

Formula: C₈H₁₂F₃NO₅
CAS Number: 1373253-22-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR