(S)-2-(4-Aminobenzamido)pentanedioic acid ≥95%

Supplier: Matrix Scientific

Synonyms: N-(4-Aminobenzoyl)-L-glutamic acid, N-(4-NH₂-Bz)-Glu-OH

075979-100G 075979-25G
102543-928EA 141.54 USD
102543-928 102551-998
(S)-2-(4-Aminobenzamido)pentanedioic acid ≥95%
(S)-2-(4-Aminobenzamido)pentanedioic acid

Hazard: Irritant

Formula: C₁₂H₁₄N₂O₅
MW: 266.2500 g/mol
MDL Number: MFCD00042821
CAS Number: 4271-30-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR