Ethyl-4,6-dichloropyrimidine-5-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Ethyl 4,6-dichloropyrimidine-5-carboxylate

075977-250MG 075977-1G
102543-916EA 238.46 USD
102543-916 102543-918
Ethyl-4,6-dichloropyrimidine-5-carboxylate ≥95%
Ethyl-4,6-dichloropyrimidine-5-carboxylate

Hazard: Irritant

Formula: C₇H₆Cl₂N₂O₂
MW: 221.0407 g/mol
MDL Number: MFCD10698803
CAS Number: 87600-72-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR