1-Benzylpiperidin-3-amine hydrochloride ≥95%
Supplier: Matrix Scientific
075973-1G
075973-5G
075973-10G
102543-896EA
355.38
USD
102543-896
102543-898
102543-900
1-Benzylpiperidin-3-amine hydrochloride ≥95%
1-Benzylpiperidin-3-amine hydrochloride
Hazard: Irritant
Formula:
C₁₂H₁₉ClN₂ |
MDL Number:
MFCD19980510 CAS Number: 368429-78-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...