Morpholin-2-ylmethanol hydrochloride ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Hydroxymethylmorpholine hydrochloride

075970-250MG 075970-1G 075970-5G
102543-878EA 498.46 USD
102543-878 102543-880 102543-882
Morpholin-2-ylmethanol hydrochloride ≥95%
Morpholin-2-ylmethanol hydrochloride

Hazard: Irritant

Formula: C₅H₁₂ClNO₂
MW: 153.61 g/mol
MDL Number: MFCD13191721
CAS Number: 144053-98-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR