2-Methoxypyrimidine-4,6-diol ≥95%

Supplier: Matrix Scientific

Synonyms: 4,6-Dihydroxy-2-methoxypyrimidine

075969-1G 075969-5G 075969-10G
102543-872EA 284.62 USD
102543-872 102543-874 102543-876
2-Methoxypyrimidine-4,6-diol ≥95%
2-Methoxypyrimidine-4,6-diol

Hazard: Irritant

Formula: C₅H₆N₂O₃
CAS Number: 1758-98-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR