3-Methoxypicolinic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 3-Methoxypyridine-2-carboxylic acid

075968-250MG 075968-1G 075968-5G
102543-866EA 400 USD
102543-866 102543-868 102543-870
3-Methoxypicolinic acid ≥95%
3-Methoxypicolinic acid

Hazard: Irritant

Formula: C₇H₇NO₃
MW: 153.1354 g/mol
MDL Number: MFCD07367569
CAS Number: 16478-52-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR