Benzyl(2-aminoethyl)(methyl)carbamate hydrochloride ≥95%

Supplier: Matrix Scientific

Synonyms: Benzyl (2-aminoethyl)(methyl)carbamate hydrochloride

075967-250MG 075967-1G
102543-862EA 456.92 USD
102543-862 102543-864
Benzyl(2-aminoethyl)(methyl)carbamate hydrochloride ≥95%
Benzyl(2-aminoethyl)(methyl)carbamate hydrochloride

Hazard: Irritant

Formula: C₁₁H₁₇ClN₂O₂
MW: 244.72 g/mol
MDL Number: MFCD11112220
CAS Number: 162576-01-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR