Methyl-2-amino-4-methylbenzoate ≥95%

Supplier: Matrix Scientific

075963-250MG 075963-1G 075963-5G
102543-838EA 427.69 USD
102543-838 102543-840 102543-842
Methyl-2-amino-4-methylbenzoate ≥95%
Methyl-2-amino-4-methylbenzoate

Hazard: Irritant

Formula: C₉H₁₁NO₂
MDL Number: MFCD09998260
CAS Number: 18595-17-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR