1-Bromo-3,5-dinitrobenzene ≥95%
Supplier: Matrix Scientific
Synonyms:
3,5-Dinitrobromobenzene
075962-5G
075962-25G
102543-834EA
113.85
USD
102543-834
102664-080
1-Bromo-3,5-dinitrobenzene ≥95%
1-Bromo-3,5-dinitrobenzene
Hazard: Irritant
Formula:
C₆H₃BrN₂O₄ MW: 247 g/mol |
MDL Number:
MFCD00156596 CAS Number: 18242-39-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...