5-(4-Chlorophenyl)-1-(2,4-dichlorophenyl)-4-methyl-N-(piperidin-1-yl)-1H-pyrazole-3-carboxamide ≥95%

Supplier: Matrix Scientific

Synonyms: Rimonabant

075960-1G 075960-5G
102543-822EA 461.54 USD
102543-822 102543-824
5-(4-Chlorophenyl)-1-(2,4-dichlorophenyl)-4-methyl-N-(piperidin-1-yl)-1H-pyrazole-3-carboxamide ≥95%
5-(4-Chlorophenyl)-1-(2,4-dichlorophenyl)-4-methyl-N-(piperidin-1-yl)-1H-pyrazole-3-carboxamide

Hazard: Irritant

Formula: C₂₂H₂₁Cl₃N₄O
CAS Number: 168273-06-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR