(S)-Morpholin-3-ylmethanol hydrochloride ≥95%

Supplier: Matrix Scientific

075957-250MG 075957-1G 075957-5G
102543-804EA 321.58 USD
102543-804 102543-806 102543-808
(S)-Morpholin-3-ylmethanol hydrochloride ≥95%
(S)-Morpholin-3-ylmethanol hydrochloride

Hazard: Irritant

Formula: C₅H₁₂ClNO₂
MW: 153.6073 g/mol
MDL Number: MFCD12547468
CAS Number: 218594-79-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR