Ethyl-2-(quinolin-2-yl)acetate ≥95%

Supplier: Matrix Scientific

Synonyms: Ethyl 2-(quinolin-2-yl)acetate

075956-250MG 075956-1G 075956-5G
102543-798EA 547.69 USD
102543-798 102543-800 102543-802
Ethyl-2-(quinolin-2-yl)acetate ≥95%
Ethyl-2-(quinolin-2-yl)acetate

Hazard: Irritant

Formula: C₁₃H₁₃NO₂
MW: 215.25 g/mol
MDL Number: MFCD03002579
CAS Number: 5100-57-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR