4-(4-Bromobenzoyl)piperidine hydrochloride ≥95%

Supplier: Matrix Scientific

Synonyms: (4-Bromophenyl)(4-piperidyl)methanone Hydrochloride

075954-1G 075954-5G
102543-790EA 640 USD
102543-790 102543-792
4-(4-Bromobenzoyl)piperidine hydrochloride ≥95%
4-(4-Bromobenzoyl)piperidine hydrochloride

Hazard: Irritant

Formula: C₁₂H₁₅BrClNO
MW: 304.6100 g/mol
MDL Number: MFCD06804519
CAS Number: 64671-00-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR