2-(4-Chloro-2-(trifluoromethyl)phenyl)acetonitrile ≥95%

Supplier: Matrix Scientific

075952-250MG 075952-1G
102543-782EA 640 USD
102543-782 102543-784
2-(4-Chloro-2-(trifluoromethyl)phenyl)acetonitrile ≥95%
2-(4-Chloro-2-(trifluoromethyl)phenyl)acetonitrile

Hazard: Irritant

Formula: C₉H₅ClF₃N
MW: 219.6 g/mol
MDL Number: MFCD04115841
CAS Number: 85386-80-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR