2-Chloro-DL-phenylalanine ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Chloro-DL-phenylalanine, 2-Amino-3-(2-chlorophenyl)propanoic acid, 2-Chlorophenylalanine

075949-5G 075949-25G
102543-768EA 236.92 USD
102543-768 102833-892
2-Chloro-DL-phenylalanine ≥95%
2-Chloro-DL-phenylalanine

Hazard: Irritant

Formula: C₉H₁₀ClNO₂
MW: 199.6300 g/mol
MDL Number: MFCD00037774
CAS Number: 14091-11-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR