1-(2-Aminoethyl)piperazine ≥95%

Supplier: Matrix Scientific

075948-10G 075948-100G
102543-762EA 36.92 USD
102543-762 102543-764
1-(2-Aminoethyl)piperazine ≥95%
1-(2-Aminoethyl)piperazine

Hazard: Irritant

Formula: C₆H₁₅N₃
MW: 129.21 g/mol
MDL Number: MFCD00005971
CAS Number: 140-31-8
UN: 2815
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR