2-Amino-2,3-dimethylbutanenitrile ≥95%

Supplier: Matrix Scientific

075947-10G 075947-100G
102543-758EA 46.15 USD
102543-758 102543-760
2-Amino-2,3-dimethylbutanenitrile ≥95%
2-Amino-2,3-dimethylbutanenitrile

Hazard: Irritant

Formula: C₆H₁₂N₂
CAS Number: 13893-53-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR