Valsartan ≥95%

Supplier: Matrix Scientific

Synonyms: (S)-2-(N-((2'-(1H-Tetrazol-5-yl)-[1,1'-biphenyl]-4-yl)methyl)pentanamido)-3-methylbutanoic acid, (S)-2-(N-((2'-(1H-Tetrazol-5-yl)[1,1'-biphenyl]-4-yl)methyl)pentanamido)-3-methylbutanoic acid

075945-5G 075945-25G
102664-076EA 146.15 USD
102664-076 102664-078
Valsartan ≥95%
Valsartan

Hazard: Irritant

Formula: C₂₄H₂₉N₅O₃
MW: 435.52 g/mol
MDL Number: MFCD00865840
CAS Number: 137862-53-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR