4-Methylbenzophenone ≥95%

Supplier: Matrix Scientific

Synonyms: p-Methylbenzophenone, Phenyl(p-tolyl)methanone

075944-10G 075944-100G
102543-746EA 15.38 USD
102543-746 102543-748
4-Methylbenzophenone ≥95%
4-Methylbenzophenone

Hazard: Irritant

Formula: C₁₄H₁₂O
MW: 196.25 g/mol
Melting Pt: 53…57 °C
MDL Number: MFCD00008553
CAS Number: 134-84-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR