6-(Trifluoromethyl)-o-anisidine ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Amino-3-methoxybenzotrifluoride, 2-Amino-3-(trifluoromethyl)anisole, 6-(Trifluoromethyl)-2-anisidine, 6-(Trifluoromethyl)-2-methoxyaniline, 2-Methoxy-6-(trifluoromethyl)aniline

075941-1G 075941-5G
102543-734EA 427.69 USD
102543-734 102543-736
6-(Trifluoromethyl)-o-anisidine ≥95%
6-(Trifluoromethyl)-o-anisidine

Hazard: Irritant

Formula: C₈H₈F₃NO
MW: 191.15 g/mol
MDL Number: MFCD09832384
CAS Number: 133863-87-3
UN: 2810
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR