Dimethyl-5-nitroisophthalate ≥95%

Supplier: Matrix Scientific

Synonyms: Dimethyl 5-nitroisophthalate, 5-Nitroisophthalic acid dimethyl ester

075940-10G 075940-100G
102543-730EA 36.92 USD
102543-730 102543-732
Dimethyl-5-nitroisophthalate ≥95%
Dimethyl-5-nitroisophthalate

Hazard: Irritant

Formula: C₁₀H₉NO₆
MW: 239.18 g/mol
MDL Number: MFCD00008429
CAS Number: 13290-96-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR