3-(2-Chlorophenyl)pyrrolidine hydrochloride ≥95%

Supplier: Matrix Scientific

075939-1G 075939-5G
102543-726EA 183.08 USD
102543-726 102543-728
3-(2-Chlorophenyl)pyrrolidine hydrochloride ≥95%
3-(2-Chlorophenyl)pyrrolidine hydrochloride

Hazard: Irritant

Formula: C₁₀H₁₃Cl₂N
MW: 218.1229 g/mol
MDL Number: MFCD03094760
CAS Number: 1095545-14-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR