Methyl-2-(3-hydroxyprop-1-yn-1-yl)benzoate ≥95%

Supplier: Matrix Scientific

075936-250MG 075936-1G
102543-714EA 684.62 USD
102543-714 102543-716
Methyl-2-(3-hydroxyprop-1-yn-1-yl)benzoate ≥95%
Methyl-2-(3-hydroxyprop-1-yn-1-yl)benzoate

Hazard: Irritant

Formula: C₁₁H₁₀O₃
CAS Number: 103606-72-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR