Ethyl-2-(5-chloro-1H-indazol-3-yl)acetate ≥95%

Supplier: Matrix Scientific

075931-1G 075931-5G
102543-690EA 569.23 USD
102543-690 102543-692
Ethyl-2-(5-chloro-1H-indazol-3-yl)acetate ≥95%
Ethyl-2-(5-chloro-1H-indazol-3-yl)acetate

Hazard: Irritant

Formula: C₁₁H₁₁ClN₂O₂
MW: 238.68 g/mol
MDL Number: MFCD00056935
CAS Number: 27512-72-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR