3,3-Dimethylbutyraldehyde ≥95%
Supplier: Matrix Scientific
Synonyms:
3,3-Dimethylbutanal
075930-5G
075930-25G
102543-686EA
100
USD
102543-686
102543-688
3,3-Dimethylbutyraldehyde ≥95%
3,3-Dimethylbutyraldehyde
Hazard: Flammable/Refrigerate
Formula:
C₆H₁₂O MW: 100.16 g/mol Boiling Pt: 104-106°C Density: 0.798 Flash Pt: 13°C |
MDL Number:
MFCD00042807 CAS Number: 2987-16-8 UN: 1989 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...