4'-Hydroxy-2'-(trifluoromethyl)acetophenone ≥95%

Supplier: Matrix Scientific

Synonyms: 1-(4-Hydroxy-2-(trifluoromethyl)phenyl)ethanone

075928-1G 075928-5G
102543-674EA 427.69 USD
102543-674 102543-676
4'-Hydroxy-2'-(trifluoromethyl)acetophenone ≥95%
4'-Hydroxy-2'-(trifluoromethyl)acetophenone

Hazard: Irritant

Formula: C₉H₇F₃O₂
MW: 204.15 g/mol
MDL Number: MFCD01091004
CAS Number: 220227-53-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR