Candesartan ≥95%

Supplier: Matrix Scientific

075926-1G 075926-5G
102543-664EA 284.62 USD
102543-664 102543-666
Candesartan ≥95%
Candesartan

Hazard: Irritant

Formula: C₂₄H₂₀N₆O₃
MDL Number: MFCD00864463
CAS Number: 139481-59-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR