7-Bromo-3,4-dihydro-2H-benzo[b][1,4]oxazine hydrochloride ≥95%

Supplier: Matrix Scientific

075924-250MG 075924-1G
102543-654EA 967.69 USD
102543-654 102543-656
7-Bromo-3,4-dihydro-2H-benzo[b][1,4]oxazine hydrochloride ≥95%
7-Bromo-3,4-dihydro-2H-benzo[b][1,4]oxazine hydrochloride

Hazard: Irritant

Formula: C₈H₉BrClNO
MW: 250.5200 g/mol
MDL Number: MFCD20278295
CAS Number: 1350738-87-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR