3-Cyclopropyl-1H-pyrazol-5-amine hydrochloride ≥95%

Supplier: Matrix Scientific

075923-1G 075923-5G
102543-650EA 640 USD
102543-650 102543-652
3-Cyclopropyl-1H-pyrazol-5-amine hydrochloride ≥95%
3-Cyclopropyl-1H-pyrazol-5-amine hydrochloride

Hazard: Irritant

Formula: C6H10ClN3
MDL Number: MFCD08702764
CAS Number: 1031791-16-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR