5-Bromo-2,3-dimethylpyridine ≥95%

Supplier: Matrix Scientific

075922-250MG 075922-1G
102543-646EA 304.62 USD
102543-646 102543-648
5-Bromo-2,3-dimethylpyridine ≥95%
5-Bromo-2,3-dimethylpyridine

Hazard: Irritant

Formula: C₇H₈BrN
MDL Number: MFCD11656270
CAS Number: 27063-90-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR