2-(Bromomethyl)-1,3-dimethylbenzene ≥95%

Supplier: Matrix Scientific

075921-1G 075921-5G
102543-642EA 156.92 USD
102543-642 102543-644
2-(Bromomethyl)-1,3-dimethylbenzene ≥95%
2-(Bromomethyl)-1,3-dimethylbenzene

Hazard: Irritant

Formula: C₉H₁₁Br
MDL Number: MFCD03844742
CAS Number: 83902-02-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR