1-((4-Chlorophenyl)(phenyl)methyl)-4-(3-methylbenzyl)piperazine dihydrochloride ≥95%

Supplier: Matrix Scientific

Synonyms: Meclizine dihydrochloride

075920-5G 075920-10G 075920-100G
102543-636EA 107.69 USD
102543-636 102543-638 102543-640
1-((4-Chlorophenyl)(phenyl)methyl)-4-(3-methylbenzyl)piperazine dihydrochloride ≥95%
1-((4-Chlorophenyl)(phenyl)methyl)-4-(3-methylbenzyl)piperazine dihydrochloride

Hazard: Irritant

Formula: C₂₅H₂₉Cl₃N₂
MW: 463.87 g/mol
MDL Number: MFCD00058199
CAS Number: 1104-22-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR