1,3-Dichloro-4-nitrobenzene ≥95%

Supplier: Matrix Scientific

075919-10G 075919-100G
102543-632EA 36.92 USD
102543-632 102543-634
1,3-Dichloro-4-nitrobenzene ≥95%
1,3-Dichloro-4-nitrobenzene

Hazard: Irritant

Formula: C₆H₃Cl₂NO₂
MDL Number: MFCD00007071
CAS Number: 611-06-3
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR